missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ AlbuMAX™ I Lipid-Rich BSA

AlbuMAX™ I Lipid-Rich BSA

Brand:  Gibco™ 11020039

 View more versions of this product

Product Code. 10114672

  • 3225.00 € / 1kg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Lipid-rich bovine serum albumin for cell culture (1). It is chromatographically purified and has an IgG content ≤ 0.1%.

Origin: New Zealand

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in refrigerator (2–8°C).
AlbuMAX™, Gibco™
Room Temperature
BSA
1 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2833722 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog Number11020039,11020088,11020021
Lot Number2494919B Certificate TypeCertificate of Analysis Date26/01/2025 Catalog Number11020039,11020088,11020021
Lot Number2769784 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog Number11020039,11020088,11020021
Lot Number2833723 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog Number11020039,11020088,11020021
Lot Number2833725 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number11020039,11020088,11020021
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ AlbuMAX™ I Lipid-Rich BSA > 1 kg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.