Learn More
Thermo Scientific™ Alkaline Phosphatase Buffer-1 (5X)
For cellular and molecular biology applications
50.65 € - 73.97 €
Specifications
Components | 200mM Tris, 200mM NaCl, and 40mM magnesium chloride at pH 9.5 |
---|---|
Content And Storage | Keep cold |
Product Type | Alkaline Phosphatase Buffer-1 (5X) |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15415209
|
Thermo Scientific™
J62907.AK |
250 mL |
73.97 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15405209
|
Thermo Scientific™
J62907.AE |
100 mL |
50.65 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Formulation: 200mM Tris, 200mM NaCl, and 40mM magnesium chloride at pH 9.5
Specifications
200mM Tris, 200mM NaCl, and 40mM magnesium chloride at pH 9.5 | |
Keep cold | |
Alkaline Phosphatase Buffer-1 (5X) | |
Liquid |
For Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.