missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan All traffic external quick-hang PVC curtain, 3000mm x 1000mm
Brand: Manutan A352098

This item is not returnable.
View return policy
Description
- Drive through flexible curtains for external doors. PVC strips create a see through flexible curtain. Keeps heat in and reduces cold air loss in chill/freezer rooms for greater temperature control. Provides protection against rain and wind. Restricts insect and bird access. Forms a good noise barrier. Full - 95% overlap. Suitable for pedestrian and heavy traffic.
- Dimensions (H x W): 3000mm x 1000mm
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Manutan All traffic external quick-hang PVC curtain, 3000mm x 1000mm
Spot an opportunity for improvement?Share a Content Correction