Learn More
B Braun™ Injekt™-F Fine Dosage Syringe
Manufactured from polypropylene/polyethylene. B. Braun™ Injekt™-F Fine Dosage Syringe has high contrast black graduation for ideal readability.
Brand:  B Braun™ 9166017V
Product Code. 10303002
  • 40.00 € / Pack of 100
Edge
Explore more special offers
Description
Description
    • Made of polypropylene/polyethylene
    • Green plunger
    • Highly transparent barrel
    • High contrast black graduation, permanent marking, ideal readability
    • Displacement spike: Reduces residual volume and minimizes unnecessary loss of medicine
    • Centric Luer cone
    • Safe plunger backstop
    • Easy aspiration up to maximum volume
    • Latex-free/PVC-free
    • Silicone oil-free
    • Single sterile-packed (in box of 100 pcs)
Specifications
Specifications
ISO 7886-1
Green Plunger
Ideal for simple injections, vaccinations and more
Centric Luer Cone 2-part Syringe
100/Pk.
Centric Luer Cone
Sterile
Polyethylene, Polypropylene
Black graduation
Yes
Single-use
1 mL
Yes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
B Braun™ Injekt™-F Fine Dosage Syringe > Volume: 1mL ; Connection type: Centric Luer-slip