missing translation for 'onlineSavingsMsg'
Learn More

Allyl isothiocyanate, 94%, stabilized with 0.01% alpha-tocopherol, Thermo Scientific Chemicals

23.50 € - 115.00 €

Chemical Identifiers

CAS 57-06-7
MDL Number MFCD00004822
InChI Key ZOJBYZNEUISWFT-UHFFFAOYSA-N
Synonym allyl isothiocyanate, mustard oil, redskin, allylsenevol, allylsenfoel, allyl mustard oil, oleum sinapis, oil of mustard, allylsevenolum, senfoel
PubChem CID 5971
ChEBI CHEBI:73224
IUPAC Name 3-isothiocyanatoprop-1-ene
SMILES C=CCN=C=S
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10224130
View Documents Promotion Details
Thermo Scientific Acros
102950050
5 g Glass bottle
23.50 €
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10284170
View Documents Promotion Details
Thermo Scientific Acros
102951000
100 g Glass bottle
35.30 €
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10264220
View Documents Promotion Details
Thermo Scientific Acros
102955000
500 g Glass bottle
115.00 €
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

57-06-7
ZOJBYZNEUISWFT-UHFFFAOYSA-N
5971
3-isothiocyanatoprop-1-ene
MFCD00004822
allyl isothiocyanate, mustard oil, redskin, allylsenevol, allylsenfoel, allyl mustard oil, oleum sinapis, oil of mustard, allylsevenolum, senfoel
CHEBI:73224
C=CCN=C=S

Specifications

57-06-7
100.0
White
151.0°C to 153.0°C
Authentic
C4H5NS
1.5290 to 1.5310
04, 214
1.0175
Solubility in water: 2g/L water (20°C). Other solubilities: ethanol of 65 vol%- 1:20 v/v,diethylphthalate - in all proportions,paraffin oil 65 cp- in all proportions,1,2-propanediol - 1:5 v/v,soluble in ether,acetone,benzene,cs2
C=CCN=C=S
0.01% alpha-tocopherol
5971
99.15
Fine Crystalline Powder
92.0
-80.0°C
1.0175g/mL
46°C
92% min. (GC)
H2C=CHCH2NCS
MFCD00004822
15, 286
allyl isothiocyanate, mustard oil, redskin, allylsenevol, allylsenfoel, allyl mustard oil, oleum sinapis, oil of mustard, allylsevenolum, senfoel
ZOJBYZNEUISWFT-UHFFFAOYSA-N
3-isothiocyanatoprop-1-ene
99.15
CHEBI:73224
94%
Allyl isothiocyanate, 94%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Very t

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remov

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 200-309-2

missing translation for 'rtecsNumber' : NX8225000

missing translation for 'tsca' : TSCA

Product Identifier
  • Allyl isothiocyanate, stabilized

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H301-Toxic if swallowed.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only