missing translation for 'onlineSavingsMsg'
Learn More

Texwipe™ AlphaSorb™ 10 Sealed-Border Polyester Wipers

For ISO Class 3 and 4 applications

Brand:  Texwipe™ TX1052

 View more versions of this product

Product Code. 18603943

  • 1140.00 € / Pack of 600

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Double-knit, no-run interlock construction of 100% continuous-filament polyester
  • Two-ply, sealed-border wiper with highest sorbency
  • Ideal wiper for use in multiple environments
  • Excellent for picking up large amounts of liquids and for applying chemicals
  • Abrasion-resistant wiper with soft, flexible hand
  • Lot-to-lot traceability

Order Info

Available dry or sterile. Basis weight: 185g/m2.

Specifications

Specifications

Cleanroom Wipe
30.5 cm
30.5 cm
185
Sealed
Ideal wipes for use in multiple environments
12 in.
12 in.
Polyester
ISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999
12 x 12 in. (30.48 x 30.48 cm)
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.