missing translation for 'onlineSavingsMsg'
Learn More
Ansell HyFlex™ 11-816 Knitted Nylon Gloves with Fortix™ Nitrile Foam Coating Product Code.: 16069671
Add to basket

Ansell HyFlex™ 11-816 Knitted Nylon Gloves with Fortix™ Nitrile Foam Coating

Product Code. 16069671 Shop All Ansell Products
6
Click to view available options
Size:
6
7
8
9
10
11
This item is not returnable. View return policy

Product Code. 16069671

Brand: Ansell 11816060

Edge
Add to basket
Add to basket
This item is not returnable. View return policy

An ultra-thin liner and coating provide a barehand feel for workers who rely on fingertip sensitivity for precision tasks. Fortix™ abrasion resistant foam nitrile coating enhances safety and productivity.

  • Ultra-thin liner and coating ensures a barehand feel for workers who rely on fingertip sensitivity for precise tasks
  • Ergonomic design provides tactility for easy handling of small parts
  • Materials are tested by dermatologists, solvent-free, and Oekotex certified for no harmful substances
  • FORTIX™ Abrasion Resistance Technology extends glove life while enhancing worker safety and productivity
  • No transfer of silicone contaminants to metal parts prior to painting
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category II
Construction Seamless Knit
Coating Material Nitrile
Color Black, Blue
Hand Specific
Latex Free No
Material Nylon, Spandex
Product Type Seamless Knit Gloves
Size 6
For Use With (Application) Automotive Assembly, Machinery/Equipment Operation, Metal Fabrication, and Transportation
Glove Cuff Style Knit
Series HyFlex™ 11-816
Yarn Gauge 18
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell HyFlex™ 11-816 Knitted Nylon Gloves with Fortix™ Nitrile Foam Coating >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.