missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Scaffolding Rods

Provides for sturdy scaffolding construction in the laboratory. Fisherbrand™ Scaffolding Rods are manufactured from a durable aluminum alloy.

Brand:  Fisherbrand™ 2000/4

 View more versions of this product

Product Code. 11567023

  • 15.15 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Approx. 13mm in diameter
  • Available in a range of lengths
  • Fits with Fisherbrand scaffolding connectors and foot adapters
Specifications

Specifications

Aluminum Alloy
900 mm
Rod
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Scaffolding Rods > Length: 900mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.