missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Scaffolding Rods
Provides for sturdy scaffolding construction in the laboratory. Fisherbrand™ Scaffolding Rods are manufactured from a durable aluminum alloy.
8.65 € - 33.90 €
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
11577023
|
Fisherbrand™
2000/3 |
1200 mm |
33.90 €
Each |
Estimated Shipment: 21-05-2025
Log in to see stock. |
||||||
11587023
|
Fisherbrand™
2000/1 |
1800 mm |
17.55 €
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||||
11557023
|
Fisherbrand™
2000/5 |
600 mm |
8.65 €
Each |
Estimated Shipment: 19-06-2025
Log in to see stock. |
||||||
11567023
|
Fisherbrand™
2000/4 |
900 mm |
15.15 €
Each |
In Stock
Log in to see stock. |
||||||
Description
- Approx. 13mm in diameter
- Available in a range of lengths
- Fits with Fisherbrand scaffolding connectors and foot adapters
Specifications
Aluminum Alloy | |
Rod |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Scaffolding Rods