missing translation for 'onlineSavingsMsg'
Learn More

R&L Enterprises™ Aluminum Alloy Rods

An excellent option for the construction of retort stands in the laboratory. R&L Enterprises™ Aluminum Alloy Rods are manufactured from aluminum alloy.

Brand:  R&L Enterprises™ 3108/42

 View more versions of this product

Product Code. 12308149

  • 2490.00 € / Pack of 2
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Threaded
  • Thread diameter 10mm
  • Length of rod does not include the thread
Specifications

Specifications

Aluminum Alloy
Aluminum Rods
2 Pk.
1000 mm
10 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
R&L Enterprises™ Aluminum Alloy Rods > Length: 1000mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.