missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ Aluminum Anodized Lab Jacks
Manufactured according to DIN 12897 standards. Bochem™ aluminum Anodized Lab Jacks features a rounded, east to use ergonomic knob.
Brand: Bochem™ 11031
This item is not returnable.
View return policy
Description
- Improved scissor technology with reduced tolerances
- Rounded, easy to use ergonomic knob
- Scissor-construction made of 18/10-stainless steel
- Maximal dynamic operating load (Kg max. dyn.) and the maximal static operating load (Kg max. stat.) norms are DIN 12897 compliant
- Type 1 (240x240mm) uses a hand-operated adjusting wheel
- Type 2 (300x300mm) includes ratchet, which connects to adjusting wheel
Specifications
Anodized Aluminum, Stainless Steel | |
200 mm | |
Wheels | |
1 Ea. |
200 x 200 mm | |
200 mm | |
Lab Jack |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bochem™ Aluminum Anodized Lab Jacks > Dimensions (L x W): 200 x 200mm
Spot an opportunity for improvement?Share a Content Correction