Learn More
Aluminum oxide, for chromatography, basic, Brockmann I, 40-300μm, 60A, Thermo Scientific Chemicals
Aluminum oxide, Al2O3, CAS Number-1344-28-1, abramant, abramax, abrarex, abrasit, aloxite, alpha-alumina, Aluminum oxide, Aluminum oxide, alundum, fasertonerde, 1kg, Na2O <0.4%, 2977.0 deg.C, White, 3.9700g/mL, 215-691-6, 300ppm max.
85.93 € - 1260.27 €
Chemical Identifiers
CAS | 1344-28-1 |
---|---|
Molecular Formula | Al2O3 |
Molecular Weight (g/mol) | 101.96 |
MDL Number | MFCD00003424 |
InChI Key | PNEYBMLMFCGWSK-UHFFFAOYSA-N |
Synonym | aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum |
PubChem CID | 9989226 |
SMILES | [O--].[O--].[O--].[Al+3].[Al+3] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10730701
|
Thermo Scientific Acros
189990010 |
1 kg | Plastic Bottle |
85.93 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10378193
|
Thermo Scientific Acros
189990051 |
5 kg | Plastic Bottle |
319.59 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10388433
|
Thermo Scientific Acros
189990250 |
25 kg | Fibre drum |
1260.27 €
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For column chromatographyChemical Identifiers
1344-28-1 | |
101.96 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
9989226 |
Al2O3 | |
MFCD00003424 | |
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundumShow More | |
[O--].[O--].[O--].[Al+3].[Al+3] |
Specifications
2977.0°C | |
1344-28-1 | |
9 to 10 (10% aq. suspension) | |
Al2O3 | |
MFCD00003424 | |
15, 350 | |
Solubility in water: insoluble. Other solubilities: slightly sol. in alkaline sol.(forming hydroxides), practically insoluble in non-polar organic solvent, difficultly soluble in mineral acids | |
[O--].[O--].[O--].[Al+3].[Al+3] | |
101.96 | |
101.96 | |
3.97 | |
Aluminum oxide |
2030.0°C | |
White | |
Powder | |
Al2O3 | |
01,19; 02,17; 03,6; 04,8; 06,16; 07,5; 08,9; 09,8; 10,8; 11,22; 13,14; 15,9; 16,9 | |
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
dialuminium(3+) trioxidandiide | |
9989226 | |
Chromatography | |
3.9700g/mL |
Safety and Handling
missing translation for 'einecsNumber' : 215-691-6
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.