Learn More
Fisherbrand™ Bossheads
For securely keeping clamps in place on laboratory scaffolding. Fisherbrand™ Bossheads come with offset jaws for greater strength.
Brand:  Fisherbrand™ 9507
 View more versions of this product
Product Code. 10758931
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • With offset knobs for greater strength
  • Aluminum alloy with clamping screws
  • For use with rods up to 16mm in diameter
  • Note: these screws have a metal head
Specifications
Specifications
5/Pk.
Aluminum Alloy
For use with rod up to 16mm diameter
Bosshead Clamp
Metal headed clamping screw
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products