missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Aluminum Alloy Labeled Safety Lockout Hasps

A combination of lockout and tagout.

Brand:  Brady™ 265393

 View more versions of this product

Product Code. 11781326

  • 146.00 € / 1 pack
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Allow a single employee to lock out and service equipment without having separate hasps and tags
  • Use permanent or non-permanent markers pens to write on the labels
Specifications

Specifications

Labeled Safety Lockout Hasps
Anodized Aluminum Alloy
8 mm
For Labeled Safety Lockout Hasps
5 Pk.
Red
4.7 mm
NE PAS FAIRE FONCTIONNER
184.1 mm
76.2 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.