Learn More
Aluminum fluoride hydrate, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals
41.60 € - 464.00 €
Chemical Identifiers
CAS | 32287-65-3 |
---|---|
Molecular Formula | AlF3H2O |
Molecular Weight (g/mol) | 101.99 |
MDL Number | MFCD00149142 |
InChI Key | XTXFDUABTPNTFB-UHFFFAOYSA-K |
Synonym | aluminum fluoride hydrate, alf3.h2o, trifluoroalumane-water 1/1, aluminum fluoride hydrate, puratronic |
PubChem CID | 16211462 |
IUPAC Name | trifluoroalumane hydrate |
SMILES | O.F[Al](F)F |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
32287-65-3 | |
101.99 | |
XTXFDUABTPNTFB-UHFFFAOYSA-K | |
16211462 | |
O.F[Al](F)F |
AlF3H2O | |
MFCD00149142 | |
aluminum fluoride hydrate, alf3.h2o, trifluoroalumane-water 1/1, aluminum fluoride hydrate, puratronic | |
trifluoroalumane hydrate |
Specifications
32287-65-3 | |
AlF3H2O | |
UN3260 | |
XTXFDUABTPNTFB-UHFFFAOYSA-K | |
trifluoroalumane hydrate | |
16211462 | |
99.99% | |
(metals basis) |
Powder | |
MFCD00149142 | |
aluminum fluoride hydrate, alf3.h2o, trifluoroalumane-water 1/1, aluminum fluoride hydrate, puratronic | |
O.F[Al](F)F | |
101.99 | |
83.98 (Anhydrous) | |
Puratronic™ | |
Aluminum fluoride hydrate |
Safety and Handling
GHS H Statement
H300-H314-H318
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
missing translation for 'tsca' : TSCA
- Aluminum fluoride hydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.