Learn More
Fisherbrand™ Aluminum Labjacks
Provides for precise height adjustment with the turn of a screw. Fisherbrand™ Aluminum Labjacks are manufactured to close tolerances with a stainless steel operating screw.
Brand:  Fisherbrand™ 2041
 View more versions of this product
Product Code. 11735396
  • 112.00 € / Each
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Blue anodized finish
  • Completely stable and free from rocking movements
  • Top and bottom plates are specially designed aluminum extrusion giving exceptional rigid and robust support
Specifications
Specifications
Aluminum
50 mm
With stainless steel operating screw
1 Ea.
36 to 78 mm
40 mm
Lab Jack