missing translation for 'onlineSavingsMsg'
Learn More

Aluminum perchlorate nonahydrate, Reagent Grade, Thermo Scientific Chemicals

53.06 € - 174.87 €

Chemical Identifiers

CAS 81029-06-3
Molecular Formula AlCl3H18O21
Molecular Weight (g/mol) 487.455
MDL Number MFCD00149130
InChI Key CYRDFMGMNQJULY-UHFFFAOYSA-K
Synonym aluminum perchlorate nonahydrate, al.3clo4.9h2o, aluminum perchlorate nonahydrate, reagent, aluminum 3+ nonahydrate triperchlorate
PubChem CID 16211522
IUPAC Name aluminum;triperchlorate;nonahydrate
SMILES O.O.O.O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Al+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11381768
View Documents
Thermo Scientific Alfa Aesar
011659.22
100 g
53.06 €
100g
Please to purchase this item. Need a web account? Register with us today!
11391768
View Documents
Thermo Scientific Alfa Aesar
011659.36
500 g
174.87 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Aluminum perchlorate nonahydrate is used as an intermediate in both pharmaceutical industry and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

81029-06-3
487.455
CYRDFMGMNQJULY-UHFFFAOYSA-K
16211522
O.O.O.O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Al+3]
AlCl3H18O21
MFCD00149130
aluminum perchlorate nonahydrate, al.3clo4.9h2o, aluminum perchlorate nonahydrate, reagent, aluminum 3+ nonahydrate triperchlorate
aluminum;triperchlorate;nonahydrate

Specifications

82°C
Crystalline
MFCD00149130
aluminum perchlorate nonahydrate, al.3clo4.9h2o, aluminum perchlorate nonahydrate, reagent, aluminum 3+ nonahydrate triperchlorate
CYRDFMGMNQJULY-UHFFFAOYSA-K
aluminum;triperchlorate;nonahydrate
16211522
Reagent
Aluminum perchlorate nonahydrate
81029-06-3
AlCl3H18O21
UN1481
Soluble in water.
O.O.O.O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Al+3]
487.455
487.46 (325.33 Anhydrous)
Hygroscopic
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H272-H314

missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 238-437-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Aluminum perchlorate nonahydrate

Signal Word
  • Danger

Hazard Category
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P220-Keep/Store away from clothing/combustible materials.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.