Learn More
R&L Enterprises™ Retort Stand Clamps
Enables articles from 2 to 90mm in diameter to be clamped securely. R&L Enterprises™ Retort Stand Clamps feature aluminum alloy jaws with other parts of plated brass.
Brand:  R&L Enterprises™ Z0104
Product Code. 11785376
  • 38.39 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Design ensures a firm grip at all angles of opening
  • Cork liners for the jaws are well secured
Specifications
Specifications
Retort Clamp
Aluminum
For use with 85mm diameter
Four Interlocking Cork Lined
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
R&L Enterprises™ Retort Stand Clamps > For Use With: For use with 85mm diameter