Learn More
Lovibond™ Comparator Disc
Lovibond™ Comparator Disc uses an array of glass fade-free color match standards for chemical test color comparison.
Brand:  Lovibond™ 230070
 View more versions of this product
Product Code. 13116040
  • 266.00 € / Each
Estimated Shipment: 20-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Lovibond™ Comparator Disc uses an array of glass fade-free color match standards for chemical test color comparison.
  • Easy to use, visual assessment of the sample; simply rotate disc to match color
  • Glass, fade-free standards for longevity of use
  • Shock resistant, scratch resistant
  • Optional lighting units for guaranteed lighting conditions 24 hours/day
  • Optional Nessleriser systems for longer path lengths
  • More than 400 color discs are available for the Comparator 2000+
  • Lovibond™ color standards consist exclusively of through-and-through colored and flat-ground glasses
Specifications
Specifications
Ammonia
Comparator 2000+
Color Comparator Disc
Disc 3/113
0, 0.1, 0.2, 0.3, 0.4, 0.5, 0.6, 0.8, 1.0 mg/L N
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lovibond™ Comparator Disc > Analyte: Ammonia