missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Silicon slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, 99.99% (metals basis), Thermo Scientific Chemicals
156.00 € - 610.04 €
Chemical Identifiers
CAS | 11145-30-5 |
---|---|
Molecular Formula | AlSi |
Molecular Weight (g/mol) | 55.067 |
MDL Number | MFCD00143595 |
InChI Key | CSDREXVUYHZDNP-UHFFFAOYSA-N |
Synonym | silicon aluminum, aluminum-silicon alloy, al.si, aluminum silicon slug, aluminum silicon, aluminum-silane 1/1, aluminum silicon powder,-325 mesh, aluminum silicon slug, 6.35mm 0.25in dia x 6.35mm 0.25in length, aluminum silicon slug, 12.7mm 0.5in dia x 12.7mm 0.5in length, puratronic, aluminum silicon slug, 6.35mm 0.25in dia x 12.7mm 0.5in length, puratronic |
PubChem CID | 9793685 |
IUPAC Name | aluminum;silicon |
SMILES | [Al].[Si] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11394378
|
Thermo Scientific Alfa Aesar
042322.18 |
50 g | N/A | ||||||
11314388
|
Thermo Scientific Alfa Aesar
042322.30 |
250 g |
610.04 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
11145-30-5 | |
55.067 | |
CSDREXVUYHZDNP-UHFFFAOYSA-N | |
9793685 | |
[Al].[Si] |
AlSi | |
MFCD00143595 | |
silicon aluminum, aluminum-silicon alloy, al.si, aluminum silicon slug, aluminum silicon, aluminum-silane 1/1, aluminum silicon powder,-325 mesh, aluminum silicon slug, 6.35mm 0.25in dia x 6.35mm 0.25in length, aluminum silicon slug, 12.7mm 0.5in dia x 12.7mm 0.5in length, puratronic, aluminum silicon slug, 6.35mm 0.25in dia x 12.7mm 0.5in length, puratronic | |
aluminum;silicon |
Specifications
11145-30-5 | |
AlSi | |
silicon aluminum, aluminum-silicon alloy, al.si, aluminum silicon slug, aluminum silicon, aluminum-silane 1/1, aluminum silicon powder,-325 mesh, aluminum silicon slug, 6.35mm 0.25in dia x 6.35mm 0.25in length, aluminum silicon slug, 12.7mm 0.5in dia x 12.7mm 0.5in length, puratronic, aluminum silicon slug, 6.35mm 0.25in dia x 12.7mm 0.5in length, puratronic | |
[Al].[Si] | |
55.067 | |
99.99% | |
Slug | |
Aluminum Silicon slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length |
Al:Si; 99:1 wt% | |
MFCD00143595 | |
CSDREXVUYHZDNP-UHFFFAOYSA-N | |
aluminum;silicon | |
9793685 | |
Odorless | |
(metals basis) |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only