Learn More
Heathrow Scientific™ Aluminum Test Tube Basket
GREENER_CHOICE
Wash and store test tubes. Heathrow Scientific™ Aluminum Test Tube Basket can withstand repeated exposure to autoclaves.
48.00 € - 2250.00 €
Specifications
Autoclavable Autoclavable
Material Aluminum
For Use With (Application) Washing, Autoclaving, Storing Test Tubes
View More Specs
Products 2
Product Code Brand Dimensions (L x W x H) Price Quantity & Availability  
Product Code Brand Dimensions (L x W x H) Price Quantity & Availability  
12317569
View Documents
Heathrow Scientific™
HS20341A
152 x 152 x 152mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12327569
GREENER_CHOICE
View Documents
Heathrow Scientific™
HS20341B
256 x 152 x 152mm
48.00 €
Each
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
Description
Description
  • Constructed of lightweight aluminum
  • Ideal for washing and storing test tubes
  • Withstands repeated autoclaving and use in the lab
Specifications
Specifications
Autoclavable
Aluminum
Washing, Autoclaving, Storing Test Tubes
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Heathrow Scientific™ Aluminum Test Tube Basket