Learn More
Heathrow Scientific™ Aluminum Test Tube Basket
Wash and store test tubes. Heathrow Scientific™ Aluminum Test Tube Basket can withstand repeated exposure to autoclaves.
Brand: Heathrow Scientific™ HS20341B
Description
- Constructed of lightweight aluminum
- Ideal for washing and storing test tubes
- Withstands repeated autoclaving and use in the lab
Specifications
Autoclavable | |
Aluminum | |
Washing, Autoclaving, Storing Test Tubes | |
256 x 152 x 152mm |