missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Aluminum Transport Signs Overlaminated with Polyester

34.70 € - 38.10 €

Specifications

Certifications/Compliance ISO 7010
Material Aluminum, Polyester
Quantity 1 Ea.
Description Aluminum with Polyester Overlaminate
For Use With (Application) Used for indoor and outdoor safety signs
View More Specs

Products 4
Product Code Brand Displayed Text Price Quantity & Availability  
Product Code Brand Displayed Text Price Quantity & Availability  
11764904
missing translation for 'viewDocuments'
Brady™
258950
Flammable Solid 4
38.10 €
Each
Estimated Shipment: 24-04-2025
to see stock.
Add to basket
 
11704914
missing translation for 'viewDocuments'
Brady™
258956
Flammable When Wet 4 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11744904
missing translation for 'viewDocuments'
Brady™
258947
Highly Flammable Liquid 3
38.10 €
Each
Estimated Shipment: 24-04-2025
to see stock.
Add to basket
 
11784904
missing translation for 'viewDocuments'
Brady™
258953
Spontaneous Ignition 4
38.10 €
Each
Estimated Shipment: 24-04-2025
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

ISO 7010
1 Ea.
Used for indoor and outdoor safety signs
0.271 mm
-40 °C-140 °C
Transport Sign
Aluminum, Polyester
Aluminum with Polyester Overlaminate
297 mm
297 mm
Diamond
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brady™ Aluminum Transport Signs Overlaminated with Polyester

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.