missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Micro Aluminum Weighing Dishes

Use these aluminum weighing dishes for microbalances, organic micro elementary and thermal analysis, and pharmaceutical applications. Micro Aluminum Weighing Dishes are compatible with most commercially available analyzers and micro balances.

Brand:  Cole-Parmer™ 01019-03

 View more versions of this product

Product Code. 11877232

  • 174.00 € / Pack of 250
Estimated Shipment: 26-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Packaged in easy-to-use plastic containers for storage and dispensing
  • Oil-free and non-coated
  • Disposable
Specifications

Specifications

Flat
Single-use
Round
Microbalances, Organic Micro Elementary and Thermal Analysis, and Pharmaceutical Use
2.5 mm
6 mm
0.035 mL
Aluminum
Weighing Dish
6 mm
0.25 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ Micro Aluminum Weighing Dishes > Capacity: 0.035mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.