Learn More
Aluminum wire, 0.5mm (0.02in) dia, hard, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Form: Approx.0.53g/m; Temper: as drawn
50.85 € - 578.88 €
Chemical Identifiers
CAS | 7429-90-5 |
---|---|
Molecular Formula | Al |
Molecular Weight (g/mol) | 26.98 |
MDL Number | MFCD00134029 |
InChI Key | XAGFODPZIPBFFR-UHFFFAOYSA-N |
Synonym | aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum |
PubChem CID | 5359268 |
ChEBI | CHEBI:33629 |
IUPAC Name | aluminium |
SMILES | [Al] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11319897
|
Thermo Scientific Alfa Aesar
042238.G6 |
10 m |
50.85 €
10m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11309897
|
Thermo Scientific Alfa Aesar
042238.H2 |
50 m |
152.76 €
50m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11399887
|
Thermo Scientific Alfa Aesar
042238.H7 |
250 m |
578.88 €
250m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7429-90-5 | |
26.98 | |
XAGFODPZIPBFFR-UHFFFAOYSA-N | |
5359268 | |
aluminium |
Al | |
MFCD00134029 | |
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum | |
CHEBI:33629 | |
[Al] |
Specifications
Silver | |
Silver-colored wire | |
Al | |
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum | |
[Al] | |
26.98 | |
CHEBI:33629 | |
Wire |
7429-90-5 | |
99.999% (Metals basis) | |
MFCD00134029 | |
XAGFODPZIPBFFR-UHFFFAOYSA-N | |
aluminium | |
5359268 | |
Odorless | |
Aluminum, Hard |
Safety and Handling
missing translation for 'einecsNumber' : 231-072-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.