missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amberlite™ IR120, H form, ion-exchange resin, Thermo Scientific™
Amberlite IR120, H form, ion-exchange resin, ACROS Organics, Quantity: 1kg, Packaging: Plastic bottle, CAS: 78922-04-0, Amber to Brown-Yellow, Molecular Formula: C13H10ClNO4S, Molecular Weight: 311.736
23.00 € - 188.10 €
Chemical Identifiers
CAS | 78922-04-0 |
---|---|
Molecular Formula | C13H10ClNO4S |
Molecular Weight (g/mol) | 311.736 |
MDL Number | MFCD00132707 |
InChI Key | APBOVLPLJFJSRI-UHFFFAOYSA-N |
Synonym | 3-3-chlorophenylsulfonamido benzoic acid, 3-3-chloro-benzenesulfonylamino-benzoic acid, 3-3-chlorophenyl sulfonyl amino benzoic acid, benzoicacid, 3-3-chlorophenyl sulfonyl amino, 3-3-chlorobenzenesulfonamido benzoic acid, amberlite ir-120, 3-3-chlorophenyl sulfonylamino benzoic acid, 3-3-chlorophenyl sulfonamido benzoic acid |
PubChem CID | 8190984 |
IUPAC Name | 3-[(3-chlorophenyl)sulfonylamino]benzoic acid |
SMILES | C1=CC(=CC(=C1)NS(=O)(=O)C2=CC(=CC=C2)Cl)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10098951
|
Thermo Scientific Acros
369722500 |
250 g | Plastic bottle | N/A | |||||
10653003
|
Thermo Scientific Acros
369720010 |
1 kg | Plastic bottle | N/A | |||||
10781604
|
Thermo Scientific Acros
369720051 |
5 kg | Plastic drum | N/A | |||||
Description
Amberlite IR120, H form, ion-exchange resin, ACROS Organics, Quantity: 1kg, Packaging: Plastic bottle, CAS: 78922-04-0, Amber to Brown-Yellow, Molecular Formula: C13H10ClNO4S, Molecular Weight: 311.736
Ion exchange resinChemical Identifiers
78922-04-0 | |
311.736 | |
APBOVLPLJFJSRI-UHFFFAOYSA-N | |
8190984 | |
C1=CC(=CC(=C1)NS(=O)(=O)C2=CC(=CC=C2)Cl)C(=O)O |
C13H10ClNO4S | |
MFCD00132707 | |
3-3-chlorophenylsulfonamido benzoic acid, 3-3-chloro-benzenesulfonylamino-benzoic acid, 3-3-chlorophenyl sulfonyl amino benzoic acid, benzoicacid, 3-3-chlorophenyl sulfonyl amino, 3-3-chlorobenzenesulfonamido benzoic acid, amberlite ir-120, 3-3-chlorophenyl sulfonylamino benzoic acid, 3-3-chlorophenyl sulfonamido benzoic acid | |
3-[(3-chlorophenyl)sulfonylamino]benzoic acid |
Specifications
Beads | |
Particle size analysis: uniformity coefficient <1.8, harmonic mean size: 0.620 to 0.830mm,% <0.300mm: <2.0%,%>1.180mm: <3.0% | |
C13H10ClNO4S | |
(H+): 53.0 to 58.0% | |
3-3-chlorophenylsulfonamido benzoic acid, 3-3-chloro-benzenesulfonylamino-benzoic acid, 3-3-chlorophenyl sulfonyl amino benzoic acid, benzoicacid, 3-3-chlorophenyl sulfonyl amino, 3-3-chlorobenzenesulfonamido benzoic acid, amberlite ir-120, 3-3-chlorophenyl sulfonylamino benzoic acid, 3-3-chlorophenyl sulfonamido benzoic acid | |
C1=CC(=CC(=C1)NS(=O)(=O)C2=CC(=CC=C2)Cl)C(=O)O | |
311.736 | |
Amberlite™ IR120 |
78922-04-0 | |
(H+):> 1.8 eq/L | |
MFCD00132707 | |
Amber to Brown/Yellow | |
APBOVLPLJFJSRI-UHFFFAOYSA-N | |
3-[(3-chlorophenyl)sulfonylamino]benzoic acid | |
8190984 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Amberlite]TS IR120 H resin
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title