missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amberlite™ IR120, H form, ion-exchange resin, Thermo Scientific™
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Description
Amberlite IR120, H form, ion-exchange resin, ACROS Organics, Quantity: 1kg, Packaging: Plastic bottle, CAS: 78922-04-0, Amber to Brown-Yellow, Molecular Formula: C13H10ClNO4S, Molecular Weight: 311.736
Ion exchange resin

Specifications
Specifications
Physical Form | Beads |
Particle Size | Particle size analysis: uniformity coefficient <1.8, harmonic mean size: 0.620 to 0.830mm,% <0.300mm: <2.0%,%>1.180mm: <3.0% |
Packaging | Plastic bottle |
Volume Total Capacity | (H+):> 1.8 eq/L |
Water Retention Capacity | (H+): 53.0 to 58.0% |
Color | Amber to Brown/Yellow |
Quantity | 1 kg |
Chemical Name or Material | Amberlite™ IR120 |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0407524 | Certificate of Analysis | 13/12/2022 | 369722500,369720010,369720051 |
A0407625 | Certificate of Analysis | 13/12/2022 | 369722500,369720010,369720051 |
A0413801 | Certificate of Analysis | 13/12/2022 | 369722500,369720010,369720051 |
A0413800 | Certificate of Analysis | 13/12/2022 | 369722500,369720010,369720051 |
A0415244 | Certificate of Analysis | 13/12/2022 | 369722500,369720010,369720051 |
Safety and Handling

Product Identifier
- Amberlite]TS IR120 H resin
- Danger
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Amberlite™ IR120, H form, ion-exchange resin, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction