Learn More
Amberlyst™ 15(H), ion exchange resin, Thermo Scientific Chemicals
Strongly acidic macroreticular resin
31.36 € - 240.20 €
Chemical Identifiers
CAS | 39389-20-3 |
---|---|
Molecular Formula | C18H18O3S |
Molecular Weight (g/mol) | 314.399 |
MDL Number | MFCD00145841 |
InChI Key | SIWVGXQOXWGJCI-UHFFFAOYSA-N |
Synonym | amberlyst 15, wet, ion exchange resin, 2-ethenylbenzenesulfonic acid; divinylbenzene, benzenesulfonic acid, ethenyl-, polymer with diethenylbenzene, ksc581g2r, amberlyst 15 ion-exchange resin, amberlite? ir120 hydrogen form, 2-ethenylbenzenesulfonic acid-1,2-diethenylbenzene 1:1, amberlite r ir120 hydrogen form, divinylbenzene-styrenesulfonic acid copolymer, 1,2-bis ethenyl benzene; 2-ethenylbenzenesulfonic acidShow More |
PubChem CID | 170197 |
IUPAC Name | 1,2-bis(ethenyl)benzene;2-ethenylbenzenesulfonic acid |
SMILES | C=CC1=CC=CC=C1C=C.C=CC1=CC=CC=C1S(=O)(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11359379
|
Thermo Scientific Alfa Aesar
089079.18 |
50 g |
31.36 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11379379
|
Thermo Scientific Alfa Aesar
089079.30 |
250 g |
87.84 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11369379
|
Thermo Scientific Alfa Aesar
089079.A1 |
1 kg |
240.20 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Amberlyst™ 15(H) is a strongly acidic cation exchange resin used as a heterogeneous acid catalysis and is suitable for non-aqueous catalysis. It is used to determine the mechanism of phenol alkylation using density functional theory (DFT). It is widely utilized as a column packing material for HPLC analysis. It is employed in separation media for Thin-layer chromatography (TLC) and paper chromatography. It is used in various chemical reactions like esterification, transesterification, Michael addition, Prins cyclization, Friedel-Crafts alkylation, acylation, cleavage of epoxides and crossed-aldol condensation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
39389-20-3 | |
314.399 | |
SIWVGXQOXWGJCI-UHFFFAOYSA-N | |
170197 | |
C=CC1=CC=CC=C1C=C.C=CC1=CC=CC=C1S(=O)(=O)O |
C18H18O3S | |
MFCD00145841 | |
amberlyst 15, wet, ion exchange resin, 2-ethenylbenzenesulfonic acid; divinylbenzene, benzenesulfonic acid, ethenyl-, polymer with diethenylbenzene, ksc581g2r, amberlyst 15 ion-exchange resin, amberlite? ir120 hydrogen form, 2-ethenylbenzenesulfonic acid-1,2-diethenylbenzene 1:1, amberlite r ir120 hydrogen form, divinylbenzene-styrenesulfonic acid copolymer, 1,2-bis ethenyl benzene; 2-ethenylbenzenesulfonic acidShow More | |
1,2-bis(ethenyl)benzene;2-ethenylbenzenesulfonic acid |
Specifications
Pale brown beads | |
Odorless | |
MFCD00145841 | |
amberlyst 15, wet, ion exchange resin, 2-ethenylbenzenesulfonic acid; divinylbenzene, benzenesulfonic acid, ethenyl-, polymer with diethenylbenzene, ksc581g2r, amberlyst 15 ion-exchange resin, amberlite? ir120 hydrogen form, 2-ethenylbenzenesulfonic acid-1,2-diethenylbenzene 1:1, amberlite r ir120 hydrogen form, divinylbenzene-styrenesulfonic acid copolymer, 1,2-bis ethenyl benzene; 2-ethenylbenzenesulfonic acid | |
SIWVGXQOXWGJCI-UHFFFAOYSA-N | |
1,2-bis(ethenyl)benzene;2-ethenylbenzenesulfonic acid | |
170197 |
39389-20-3 | |
C18H18O3S | |
Brown | |
Insoluble in water. | |
C=CC1=CC=CC=C1C=C.C=CC1=CC=CC=C1S(=O)(=O)O | |
314.399 | |
Amberlyst™ 15(H) |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.