Learn More
Invitrogen™ Cells-to-CT™ Bulk Lysis Reagents

Cells-to-CT™ Bulk Lysis Reagents are components of the TaqMan™ Gene Expression Cells-to-CT Kit, made available as a separate purchase, in larger volumes than provided with the kit.
Brand: Invitrogen™ 4391851C
10673.81 EUR valid until 2025-12-31
Use promo code "24340" to get your promotional price.
Includes: Lysis Solution (124mL), Stop Solution (12.5mL), DNase (1.25mL)
Description
Cells-to-CT™ Bulk Lysis Reagents are components of the TaqMan™ Gene Expression Cells-to-CT Kit, made available as a separate purchase, in larger volumes than provided with the kit. The reagents included are: Lysis Solution, Stop Solution, and DNase. The TaqMan Gene Expression Cells-to-CT Kit enables researchers to easily perform expression analysis using cultured cells without purification of RNA.
Order Info
Shipping Condition: Dry ice

Specifications
This kit contains the following: |
|
2500 reactions | |
Cell Lysis Buffer | |
Dry Ice |
Fewer resources used and less waste, Less hazardous | |
Cells-to-CT™ | |
2,500 reactions |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2829691 | Certificate of Analysis | 29/01/2025 | 4391851C |
2818853 | Certificate of Analysis | 29/01/2025 | 4391851C |
2829169 | Certificate of Analysis | 29/01/2025 | 4391851C |
2840112 | Certificate of Analysis | 29/01/2025 | 4391851C |
2855070 | Certificate of Analysis | 28/01/2025 | 4391851C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.