missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNaseOUT™ Recombinant Ribonuclease Inhibitor

Protects mRNA and improves total cDNA yields

Brand:  Invitrogen™ 10777019

261.30 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.


Product Code. 10154652

  • 335.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

RNaseOUT™ Recombinant Ribonuclease Inhibitor is a potent noncompetitive inhibitor of pancreatic-type ribonucleases such as RNase A, and is used to avoid RNA degradation in a variety of applications. RNaseOUT™ Recombinant Ribonuclease Inhibitor is an acidic protein with a molecular weight of ∼52 kDa. RNaseOUT™ inhibits RNase A, RNase B, and RNase C.

Applications

cDNA synthesis, RT-PCR, and in vitro transcription and translation

Source

Purified by affinity chromatography from E. coli expressing a cloned porcine gene

Performance and quality testing

SDS-PAGE purity, endodeoxyribonuclease assay, protein concentration, specific activity, performance evaluated by RT-PCR

Unit definition

One unit inhibits 5 ng of RNase A by 50% using cytidine 2', 3' cyclic monophosphate (cCMP) as the substrate

Unit reaction conditions

100 mM Tris-acetate (pH 6.5), 1 mM EDTA, 0.2 mM cCMP, 2 μg RNase A in 1 mL from 0 to 10 min at 25°C

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

40 U/μL
10 min.
5000 units
Store at -20°C. Avoid exposing product to frequent changes in temperature. RNaseOUT™ Ribonuclease Inhibitor requires 1 mM DTT to maintain activity.

The product is guaranteed for 6 months from date of purchase unless otherwise stated in the product documentation.
RNaseOUT™
Dry Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3037375 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number10777019,10777019_LITL18 PROMO
Lot Number2505363 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number10777019,10777019_LITL18 PROMO
Lot Number2478350 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number10777019,10777019_LITL18 PROMO
Lot Number2489605 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number10777019,10777019_LITL18 PROMO
Lot Number2475863 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number10777019,10777019_LITL18 PROMO
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ RNaseOUT™ Recombinant Ribonuclease Inhibitor > 5000 units

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.