missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cells-to-cDNA™ II Cell Lysis Buffer (10 ml Bottle)

Product Code. 10696355
Click to view available options
Quantity:
5 x 10 mL bottle
Unit Size:
50mL
This item is not returnable. View return policy

Product Code. 10696355

Brand: Invitrogen™ AM8723

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The same lysis buffer found in the Invitrogen Cells-to-cDNA™ II Kit. Supplied in five 10 mL bottles.

The same lysis buffer found in the Invitrogen Cells-to-cDNA™ II Kit. Supplied in five 10 mL bottles.

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C).
Product Line Ambion, Cells-to-cDNA
Product Type Cell Lysis Buffer
Quantity 5 x 10 mL bottle
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.