missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Single Cell Lysis Kit
GREENER_CHOICE

Product Code. 12033765
Click to view available options
Quantity:
100 preps
Unit Size:
100 reactions
This item is not returnable. View return policy

Product Code. 12033765

Brand: Invitrogen™ 4458235

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

100μL Single Cell Dnase I, 1mL Single Cell Lysis Solution, 100μL Single Cell Stop Solution

Single-cell analysis has the potential to transform life science research. However, detecting and quantitating the minute amounts of RNA contained in single cells remains a significant challenge.

Single-cell analysis has the potential to transform life science research. However, detecting and quantitating the minute amounts of RNA contained in single cells remains a significant challenge.

  • Optimized sample prep—Enables RNA preparation for single cells
  • Efficient design—No sample loss ensures maximal sensitivity
  • Versatile application—Compatible with common enzymatic reactions

Optimized RNA Sample Prep for 1–10 Cells
The Invitrogen Single Cell Lysis Kit enables RNA preparation from 1 to 10 cells. The Single Cell Lysis kit generates a concentrated cell lysate through the optimized combination of lysis buffer, stop solution, and DNase. The resulting lysate is compatible with common enzymatic reactions such as reverse transcription. Sample preparation is fast and can be completed in only 7 minutes with two pipetting steps.

No Sample Loss Ensures Maximal Sensitivity
The Invitrogen Single Cell Lysis Kit bypasses the need for traditional RNA purification. There are no sample transfers, columns or washes which can lead to loss and/or dilution of the precious sample. The entire lysate can be used in downstream reactions ensuring maximal sensitivity and accuracy in your experiments.

Compatible with Common Enzymatic Reactions
The reagents in the Invitrogen Single Cell Lysis Kit are identical to the lysis reagents in the complete Single Cell-to-CT™ Kit for Real-time RT-PCR analysis from 1 to 10 cells. They have been validated with multiple reverse transcription kits including SuperScript™ VILO™ cDNA Synthesis Kit, High Capacity RNA-to-cDNA kit and TaqMan™ MicroRNA Reverse Transcription Kit, for analysis of mRNA as well as miRNA targets. As a standalone module, the Single Cell Lysis Kit allows you to create new workflows to expand the possibilities of single cell analysis. 100 preps.

Not high-throughput compatible (Manual)

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Kit contains:
  • 100 μL Single Cell Dnase I: -20°C;
  • 1 mL Single Cell Lysis Solution: 4°C;
  • 100 μL Single Cell Stop Solution: -20°C
Green Features Fewer resources used and less waste, Less hazardous
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line Ambion, Cells-to-CT
Purification Time 7 min.
Quantity 100 preps
Shipping Condition Dry Ice
Target Lysate, ready for real-time PCR
For Use With (Application) Real Time PCR (qPCR), RT-PCR
Final Product Type Lysate
No. of Reactions 100 preps
Sample Type Bacteria, Blood, Cells, Tissue, Viral Samples
Starting Material 1-10 cells
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.