missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNA-free™ DNA Removal Kit

Safely eliminate DNA contamination from RNA samples

Brand:  Invitrogen™ AM1906

155.63 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 10729525

  • 201.00 € / 50 reactions

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

DNA-free™ DNase treatment and removal reagents are designed for removal of contaminating DNA from RNA samples and for removal of DNase after treatment.

  • No organic extraction or heat inactivation required
  • Includes novel reagent to remove DNase
  • Recombinant DNase I is certified RNase-freeThe DNA-free

  • Inactivation and removal of DNase
  • DNA-free reagents effectively remove DNase and divalent cations from reaction mixture
  • DNase/cation removal step takes only three minutes
  • No organic extraction, EDTA addition, or heat inactivation is required

TURBO™ DNA-free™ Kit is similar to the DNA-free Kit but includes TURBO DNase, an engineered hyperactive DNase that exhibits up to 350% greater catalytic efficiency than wild type DNase. The enzyme also has a 6-fold lower Km for DNA, thus enabling effective removal of trace quantities of DNA contamination.

PCR & Real-Time PCR, Real Time PCR (qPCR), Reverse Transcription

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store rDNase1 , 10X DNase 1 Buffer and DNase Inactivation Reagent at -20°C. Store Nuclease–free Water at room temperature.
DNase
50 reactions
Ambion™, DNA-free
Dry Ice
10X Reaction Buffer
DNA Removal Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2820316 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM1906
Lot Number2814585 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM1906
Lot Number2839386 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM1906
Lot Number2848460 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberAM1906
Lot Number2850462 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberAM1906
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ DNA-free™ DNA Removal Kit > 50 reactions

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.