missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RIPA Lysis and Extraction Buffer

Lyse cultured mammalian cells with this high-quality, ready-to-use and fully disclosed formulation of a popular cell lysis reagent.

Brand:  Thermo Scientific™ 89900

105.45 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.


 View more versions of this product

Product Code. 10017003

  • 114.00 € / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ RIPA Buffer is a high-quality, ready-to-use and fully disclosed formulation of a popular cell lysis reagent for cultured mammalian cells.

This RIPA buffer effectively lyses and extracts protein from cultured mammalian cells, including plated cells and pelleted suspension cells. The popular reagent enables the extraction of membrane, nuclear and cytoplasmic proteins and is compatible with many applications, including reporter assays, the Thermo Scientific BCA Protein Assay, immunoassays and protein purification. Inhibitors such as Thermo Scientific Halt Protease Inhibitor Cocktail (Part No. 78430) and Halt Phosphatase Inhibitor Cocktail (Part No. 78420) are also compatible with this RIPA buffer formulation and can be added before use to prevent proteolysis and maintain protein phosphorylation.

Highlights:

  • Convenient – ready-to-use solution

  • Flexible – compatible with many applications, including reporter assays, protein assays, immunoassays and protein purification

  • Versatile – enables extraction of cytoplasmic, membrane and nuclear proteins

  • Disclosed formulation – contains no proprietary components, providing users with complete control and knowledge of possible compatibility issues

TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 mL
Extraction Buffer
Upon receipt store at 4°C.
Liquid
100 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberAA402189 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number89900
Lot NumberZK398783 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number89900
Lot NumberZK399394 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number89900
Lot NumberZK400575 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number89900
Lot NumberZI397900 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number89900
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ RIPA Lysis and Extraction Buffer > 100mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.