Learn More
Invitrogen™ DNAZap™ PCR DNA Degradation Solutions
DNAZap™ Solutions are two solutions that are innocuous when used alone, but become a potent nucleic acid degrading reagent when mixed.
Brand: Invitrogen™ AM9890
Description
DNAZap™ Solutions are two solutions that are innocuous when used alone, but become a potent nucleic acid degrading reagent when mixed. This mixture is able to instantaneously degrade high levels of contaminating DNA and RNA from surfaces. Features of DNAZap™ Solutions:
- Completely degrades contaminating DNA and RNA at the level of PCR sensitivity
- Works on contact
- Degrades nucleic acid, unlike other products on the market that only act as detergents
- Ideal for cleaning PCR tubes, PCR machine surfaces, pipettors, lab benches, lab equipment, and microfuge tubes

Specifications
Liquid | |
Room Temperature | |
DNA Decontamination Reagent |
DNAZap™ Solution 1 and DNAZap™ Solution 2 should be stored at 2–8°C. | |
250 mL | |
Ambion™, DNAZap™ |
Safety and Handling
- DNAZAP 2 X 250ML EA bottle
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Copper (II) sulphatepentahydrate, 2-methoxyethanol
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.