Learn More
Invitrogen™ Poly(A)Purist™ MAG Kit
Ambion™ Poly(A)Purist™ MAG Kit (patent pending) uses magnetic technology for the selection of mRNA that typically requires only a single round of selection yet yields RNA that is more highly enriched and devoid of rRNA than other procedures.
Brand: Invitrogen™ AM1922M
872.00 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Includes: Store Nuclease–free Water, Microfuge Tubes and Microfuge tubes with Spin Columns at room temperature. Store 2X Binding Solution, Wash Solution 1, Wash Solution 2 and Oligo(dT) Cellulose at 4°C. Store RNA Storage Solution, 5M Ammonium Acetate and Glycogen(5mg/mL) at -20°C.
Requires : Magnetic stand
This item is not returnable. View return policy
Description
Ambion™ Poly(A)Purist™ MAG Kit (patent pending) uses magnetic technology for the selection of mRNA that typically requires only a single round of selection yet yields RNA that is more highly enriched and devoid of rRNA than other procedures. Each kit contains sufficient reagents for the isolation of mRNA from up to 8 mg of total RNA.
• Highest recovery of mRNA of any available kit
• High stringency for lowering residual rRNA content
• Single round of selection sufficient for most applications
The superior performance of Poly(A)Purist™ MAG Kit is clearly demonstrated by a comparison of the mRNA recovered using 4 other kits (see Figure). The kit uses batch binding of RNA to premeasured aliquots of oligo(dT) cellulose to circumvent the problems of slow flow rates and clogged columns often experienced during conventional, gravity-driven chromatography. The magnetic bead–based selection employed in the delivers increased processing speed, ease of handling, and scalability. The entire selection procedure can be completed in as little as 45 minutes (see Figure).
Accessory Products:
A magnetic stand is required for use this kit. They are available in several formats, including the Single Place Magnetic Stand (Cat. No. AM10026) and the 6 Tube Magnetic Stand (Cat. No. AM10055).
User Guide Alert (July 2023): Step 3 of the Preparation of Oligo (dT) MagBeads section (page 7) of the User Guide is misleading. Please follow this instruction for Step 3: add an equivalent mass of Oligo (dT) Magbeads into the selected empty tube and then continue with Oligo (dT) Magbeads preparation according to steps 4-9.
Order Info
Shipping Condition: Room Temperature
Specifications
Store Nuclease-free Water, Microfuge Tubes and Microfuge tubes with Spin Columns at room temperature. Store 2X Binding Solution, Wash Solution 1, Wash Solution 2 and Oligo(dT) Cellulose at 4°C. Store RNA Storage Solution, 5M Ammonium Acetate and Glycogen(5 mg/mL) at -20°C. | |
Magnetic Bead, Oligo dT Affinity | |
mRNA | |
Microarray Analysis, Reverse Transcriptase PCR (RT-PCR), Microinjection, Nucleic Acid Labeling, In Vitro Translation, cDNA Library Construction, Northern Blotting, Nuclease Protection Assays | |
MAG Kit with Manual | |
mRNA |
Kit | |
RNA | |
Not High-throughput Compatible (Manual) | |
Ambion™, Poly(A)Purist™ | |
1 kit | |
45 min. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.