Learn More
Invitrogen™ RNaseAlert™ Lab Test Kit
Invitrogen™ RNaseAlert™ Lab Test kit is a Patent-pending technology detects RNase activity in a convenient and sensitive assay that delivers results in real time.
Brand: Invitrogen™ AM1964
Description
Invitrogen™ RNaseAlert™ Lab Test kit is a Patent-pending technology detects RNase activity in a convenient and sensitive assay that delivers results in real time. Suitable for testing small sample numbers and can be used to ensure that solutions, tubes, tips, etc. are RNase-free; the kit contains sufficient reagents for 25 reactions.
- Detects as little as 3.5 x 10-7 units (∼0.5 pg) of RNase A, with a fluorometer or by eye
- Simple, straightforward 30 min. assay
- Monitor RNase activity in column fractions during protein purification
- Use with DNaseAlert™ to quantitatively detect both RNases and DNases in the same sample
Specifications
Lyophilized | |
Dry Ice | |
RNase Control Reagent |
Store Florescent Substrate, 10X RNaseAlert Lab Test Buffer, and RNase A at -20°C. Store RnaseZap™ and nuclease-free water at room temperature. |
|
25 reactions | |
Ambion™, RNaseAlert™ |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.