missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNaseAlert™ QC System

Invitrogen™ RNaseAlert™ Lab Test kit is a Patent-pending technology detects RNase activity in a convenient and sensitive assay that delivers results in real time.

Brand:  Invitrogen™ AM1966


Product Code. 10524065

  • 1384.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Invitrogen™ RNaseAlert™ Lab Test kit is a Patent-pending technology detects RNase activity in a convenient and sensitive assay that delivers results in real time. Suitable for testing small sample numbers and can be used to ensure that solutions, tubes, tips, etc. are RNase-free; the kit contains sufficient reagents for 25 reactions.

  • Detects as little as 3.5 x 10-7 units (∼0.5 pg) of RNase A, with a fluorometer or by eye
  • Simple, straightforward 30 min. assay
  • Monitor RNase activity in column fractions during protein purification
  • Use with DNaseAlert™ to quantitatively detect both RNases and DNases in the same sample
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Lyophilized
Dry Ice
RNase Control Reagent

Store RnaseAlert Substrate, 10X Rnase Alert Buffer, Rnase A, and TE Buffer pH 7.0 at -20°C.

Store RNaseZap™ Solution and Nuclease-free Water at room temperature.

480 Assays
Ambion™, RNaseAlert™
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2819911 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM1966
Lot Number2806406 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM1966
Lot Number2797969 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM1966
Lot Number2838071 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM1966
Lot Number2848424 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberAM1966
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ RNaseAlert™ QC System > 480 assays

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.