missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TE, pH 7.0, RNase-free

Invitrogen TE buffer is a commonly recommended storage solution for RNA samples. 10 mM Tris-HCl, 1 mM EDTA, pH 7.0 is in nuclease-free, ultrapure water.

Brand:  Invitrogen™ AM9861

57.58 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10490975

  • 82.25 € / 50mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Invitrogen TE buffer is a commonly recommended storage solution for RNA samples. 10 mM Tris-HCl, 1 mM EDTA, pH 7.0 is in nuclease-free, ultrapure water. The last step in every RNA isolation protocol is to resuspend the purified RNA pellet. After painstakingly isolating the RNA, it is crucial that the pellet be suspended and stored in a safe, RNase-free solution. TE Buffer is prepared from the purest reagents available, under scrupulously monitored conditions in a state-of-the-art manufacturing facility. It is then subjected to rigorous quality control procedures to guarantee their purity and performance.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

7.0
Ambion
Bottle
Liquid
50 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.