Learn More
Invitrogen™ UltraPure™ BSA (50 mg/mL)
Ambion Ultrapure BSA is a 'non-acetylated' BSA pure enough to use when DNA or RNA integrity is critical.
243.00 € - 828.00 €
Specifications
Content And Storage | Store at –20°C. |
---|---|
Concentration | 50 mg/mL |
Product Line | Ambion™ |
Shipping Condition | Dry Ice |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10743447
|
Invitrogen™
AM2616 |
1 x 50 mg |
243.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10536735
|
Invitrogen™
AM2618 |
5 x 50 mg |
828.00 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ambion Ultrapure BSA is a 'non-acetylated' BSA pure enough to use when DNA or RNA integrity is critical.
• Tubes containing 50 mg are provided, at a concentration of 50 mg/mL
• Bovine Serum Albumin (BSA) has many uses as a carrier protein and as a stabilizing agent in enzymatic reactions
• In northern, Southern, and dot blot hybridizations, BSA is also used as a blocking agent
• It is recommended in buffers for nick translation, polymerase reactions, and ligations
BSA is also a common additive for PCR amplifications, footprinting, and gel shift assays. In restriction digests, BSA has been shown to enhance enzyme activity. The molecular weight is 68 kD. Ultrapure BSA is rigorously tested for DNase, RNase, endonuclease, protease, peroxidase, and alkaline phosphatase activity, and assayed for fluorescence background. Exonuclease Activity: 3' - 5'.
Order Info
Shipping Condition: Dry Ice
Specifications
Store at –20°C. | |
Ambion™ | |
Liquid |
50 mg/mL | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.