missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ BSA (50 mg/mL)

Ambion Ultrapure BSA is a 'non-acetylated' BSA pure enough to use when DNA or RNA integrity is critical.

Brand:  Invitrogen™ AM2616

 View more versions of this product

Product Code. 10743447

  • 243.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Ambion Ultrapure BSA is a 'non-acetylated' BSA pure enough to use when DNA or RNA integrity is critical.

• Tubes containing 50 mg are provided, at a concentration of 50 mg/mL
• Bovine Serum Albumin (BSA) has many uses as a carrier protein and as a stabilizing agent in enzymatic reactions
• In northern, Southern, and dot blot hybridizations, BSA is also used as a blocking agent
• It is recommended in buffers for nick translation, polymerase reactions, and ligations

BSA is also a common additive for PCR amplifications, footprinting, and gel shift assays. In restriction digests, BSA has been shown to enhance enzyme activity. The molecular weight is 68 kD. Ultrapure BSA is rigorously tested for DNase, RNase, endonuclease, protease, peroxidase, and alkaline phosphatase activity, and assayed for fluorescence background. Exonuclease Activity: 3' - 5'.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at –20°C.
Ambion™
Dry Ice
50 mg/mL
1 x 50 mg
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2789165 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM2616
Lot Number2797853 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM2616
Lot Number2779234 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM2616
Lot Number2817018 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM2616
Lot Number2773312 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM2616
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ UltraPure™ BSA (50 mg/mL) > 1 x 50mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.