missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ ECL™ Direct Nucleic Acid Labeling/Detection System

Use for fast and easy detection of medium-to-high-target amounts. Cytiva Life Sciences™ Amersham™ ECL™ Direct Nucleic Acid Labeling/Detection System are based on the direct labeling of DNA or RNA probes with horseradish peroxidase (HRP) in a simple 20 min chemical reaction

Brand:  Cytiva RPN3006


 View more versions of this product

Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 10459774

  • 1628.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: 45-000-922 and 45-000-923: Labeling reagent; Crosslinker; Control DNA; Blocking agent; ECL detection reagents and ECL Gold hybridization buffer
This item is not returnable. View return policy

Description

Description

 

  • Fast, easy detection of medium- to high-target amounts in 4 hr.
  • Sufficient for hybridization of 4000cm2

 

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ECL™ Direct Nucleic Acid Labeling and Detection System
ECL Gold Hybridization Buffer sufficient for 4000 cm²
ECL Gold Hybridization Buffer
Sufficient for 4000 cm2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • ACUTE AQUATIC Acute 1

Hazard Statement
  • H400-Very toxic to aquatic life.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : urea, sodium dodecylsulphate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.