missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ ECL™ Plex Fluorescent Rainbow Markers

Optimized for use with the ECL Plex Western Blotting Detection System. Amersham™ ECL™ Plex Fluorescent Rainbow Markers are ideal for quantitative western blot.

Brand:  Cytiva RPN850E


 View more versions of this product

Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 10320125

  • 119.87 € / 120µL
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Monitor progress of protein electrophoresis and assess transfer efficiency and molecular weight of blotted proteins without staining
  • Enhanced to enable faster and simpler identification of proteins on SDS-polyacrylamide gels
  • Ready-to-load
  • Provide sharper, more intense bands on gels and blots and have improved band spacing for more accurate molecular weight determination
  • Provides fluorescent images using Cy3 and Cy5 channels

Order Info

Each 120μL pack size is sufficient for use with at least 40 minigels (10 x 8cm) or 20 large gels (20 x 20cm). Each 500μL pack size is sufficient for use with at least 160 minigels (10 x 8cm) or 80 large gels (20 x 20cm).

TRUSTED_SUSTAINABILITY
Specifications

Specifications

These fluorescent rainbow markers are for use with the ECL Plex Western blotting detection system.
Protein Molecular Weight Marker/Ladder
For quantitative western blots
-20°C
Multi
Supplied in 35% Glycerol and sample buffer containing Mercaptoethanesulphonic Acid (MESNA) as reducing agent
120 μL
12 Minigel Loadings
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash hands thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : sodium dodecyl sulphate, mesna, Glycine,N,N'-1, 2-ethanediylbis¢N-(carboxymethyl)-, sodium salt, hy
  • UNKNOWN ACUTE-35.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 32.5% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.