missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ ECL™ Plex Fluorescent Rainbow Markers
Optimized for use with the ECL Plex Western Blotting Detection System. Amersham™ ECL™ Plex Fluorescent Rainbow Markers are ideal for quantitative western blot.
116.49 € - 341.00 €
Specifications
Description | These fluorescent rainbow markers are for use with the ECL Plex Western blotting detection system. |
---|---|
Color | Multi |
Product Type | Protein Molecular Weight Marker/Ladder |
Formulation | Supplied in 35% Glycerol and sample buffer containing Mercaptoethanesulphonic Acid (MESNA) as reducing agent |
For Use With (Application) | For quantitative western blots |
Product Code | Brand | Quantity | Formulation | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Formulation | Price | Quantity & Availability | ||||
10320125
|
Cytiva
RPN850E |
120 μL | Supplied in 35% Glycerol and sample buffer containing Mercaptoethanesulphonic Acid (MESNA) as reducing agent |
116.49 €
120µL |
Estimated Shipment: 02-10-2024
Log in to see stock. |
||||
10095554
|
Cytiva
RPN851E |
500 μL | Supplied in 35% Glycerol and sample buffer containing Mercaptoethanesulphonic Acid (MESNA) as reducing agent |
341.00 €
500µL |
Estimated Shipment: 01-10-2024
Log in to see stock. |
||||
Description
- Monitor progress of protein electrophoresis and assess transfer efficiency and molecular weight of blotted proteins without staining
- Enhanced to enable faster and simpler identification of proteins on SDS-polyacrylamide gels
- Ready-to-load
- Provide sharper, more intense bands on gels and blots and have improved band spacing for more accurate molecular weight determination
- Provides fluorescent images using Cy3 and Cy5 channels
Order Info
Each 120μL pack size is sufficient for use with at least 40 minigels (10 x 8cm) or 20 large gels (20 x 20cm). Each 500μL pack size is sufficient for use with at least 160 minigels (10 x 8cm) or 80 large gels (20 x 20cm).
Specifications
These fluorescent rainbow markers are for use with the ECL Plex Western blotting detection system. | |
Protein Molecular Weight Marker/Ladder | |
For quantitative western blots |
Multi | |
Supplied in 35% Glycerol and sample buffer containing Mercaptoethanesulphonic Acid (MESNA) as reducing agent | |
-20°C |
Safety and Handling
missing translation for 'shelfLife' : 3 Months
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash hands thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : sodium dodecyl sulphate, mesna, Glycine,N,N'-1, 2-ethanediylbis¢N-(carboxymethyl)-, sodium salt, hy
- UNKNOWN ACUTE-35.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 32.5% of components with unknown hazards to the aquatic environment.