missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ ECL Plex™ Western Blotting Combination Packs

Complete system optimized for fluorescent Western blotting

Brand:  Cytiva RPN999


 View more versions of this product

Product Code. 10449774

  • 304.00 € / 1 set
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket

Includes:

Amersham ECL Plex goat-α-mouse IgG-Cy3, 150μg; Amersham ECL Plex goat-α-rabbit IgG-Cy5, 150μg; Amersham ECL Plex Fluorescent Rainbow Markers, 120μL; 45-001-214: Amersham Protran Premium, 10 x 10cm, 10 sheets or 45-001-215: Hybond LFP 0.2μM PVDF, 20 x 20cm, 3 sheets

This item is not returnable. View return policy

Description

Description

  • Improved formulation of Cy3 conjugates for even lower background

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Western Blot Kit
ECL Plex western blotting system
1 Kit: ECL Plex Goat-α-mouse IgG-Cy3: 150 μg, ECL Plex Goat-α-rabbit IgG-Cy5: 150 μg, ECL Plex Fluorescent Rainbow Markers: 120 μL, Hybond LFP: 10 Sheets (20 x 20cm)
Contains Cy3, Cy5, Amersham Hybond LFP 0.2 PVDF
ECL Plex Goat-α-mouse IgG-Cy3 150μg
ECL Plex Goat-α-rabbit IgG-Cy5 150μg
ECL Plex Fluorescent Rainbow Markers 120μL
Hybond LFP, 10 Sheets (20 x 20cm)
Varies by component
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1

Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash hands thoroughly after handling.
  • P273-Avoid release to the environment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : cy3 (mono reactive),cy5 (mono reactive), sodium azide, glycerol, sodium dodecylsulphate, mensa

SDS
Documents

Documents

Certificates
Special Offers

Special Offers