missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ ECL Plex™ Western Blotting Combination Packs

Complete system optimized for fluorescent Western blotting

304.00 € - 330.00 €

Specifications

Product Type Western Blot Kit
For Use With (Application) ECL Plex western blotting system
Storage Requirements Varies by component
View More Specs

Includes: Amersham ECL Plex goat-α-mouse IgG-Cy3, 150μg; Amersham ECL Plex goat-α-rabbit IgG-Cy5, 150μg; Amersham ECL Plex Fluorescent Rainbow Markers, 120μL; 45-001-214: Amersham Protran Premium, 10 × 10cm, 10 sheets or 45-001-215: Hybond LFP 0.2μM PVDF, 20 × 20cm, 3 sheets

Products 2
Product Code Brand Formulation Price Quantity & Availability  
Product Code Brand Formulation Price Quantity & Availability  
10585545
View Documents
Cytiva
RPN998
Contains Cy3, Cy5, Amersham Protran Premium 0.45 NC
330.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
10449774
View Documents
Cytiva
RPN999
Contains Cy3, Cy5, Amersham Hybond LFP 0.2 PVDF
304.00 €
1 set
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
 
Description

Description

  • Improved formulation of Cy3 conjugates for even lower background

Specifications

Specifications

Western Blot Kit
ECL Plex western blotting system
Varies by component
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235-Keep cool.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

Supplemental information
  • MIXTURE LIST-Contains : cy5 (mono-reactive),sodium azide, nitrocellulose, glycerol,sodium dodecyl sulphate, cy3 (mono-reac

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.