Learn More
Cytiva Amersham™ ECL™ Western Blotting Detection Reagents
Use for all routine Western blotting. Cytiva Life Sciences™ Amersham™ ECL™ Western Blotting Detection Reagents are the world's first commercially available chemiluminescent detection reagents and still one of the most widely used.
Brand: Cytiva RPN2209
Description
- Detectable on Hyperfilm™ ECL or with CCD imager
- Uses enhanced luminol-based detection which is suitable for all routine Western Blotting
Western Blotting
Specifications
ECL Western Blot Detection Reagent | |
125 mL | |
Detection of 2000cm2 of membrane | |
2°C to 8°C |
Western Blot | |
∼10 pg | |
125mL detection reagent, 1125mL detection reagent 2 |