missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ ECL™ Western Blotting Detection Reagents
Use for all routine Western blotting. Cytiva Life Sciences™ Amersham™ ECL™ Western Blotting Detection Reagents are the world's first commercially available chemiluminescent detection reagents and still one of the most widely used.
206.00 € - 413.00 €
Specifications
Product Type | ECL Western Blot Detection Reagent |
---|---|
For Use With (Application) | Western Blot |
Sensitivity | ∼10 pg |
Content And Storage | 2°C to 8°C |
Product Code | Brand | Quantity | Kit Contents | Sufficient For | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Kit Contents | Sufficient For | Price | Quantity & Availability | |||
10754557
|
Cytiva
RPN2109 |
62.5 mL | 62.5mL detection reagent, 162.5mL detection reagent 2 | Detection of 1000cm2 of membrane |
206.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10340125
|
Cytiva
RPN2209 |
125 mL | 125mL detection reagent, 1125mL detection reagent 2 | Detection of 2000cm2 of membrane |
321.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10155854
|
Cytiva
RPN2106 |
250 mL | 250mL detection reagent, 1250mL detection reagent 2 | Detection of 4000cm2 of membrane |
413.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Detectable on Hyperfilm™ ECL or with CCD imager
- Uses enhanced luminol-based detection which is suitable for all routine Western Blotting
Western Blotting
Specifications
ECL Western Blot Detection Reagent | |
Western Blot | |
∼10 pg | |
2°C to 8°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva Amersham™ ECL™ Western Blotting Detection Reagents