missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Rolls
Use for the best results with fluorescence Western blotting detection applications. Cytiva Life Sciences™ Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.
Brand: Cytiva 10600009
287.89 EUR valid until 2025-12-31
Use promo code "24363" to get your promotional price.
This item is not returnable.
View return policy
Description
Supplied in rolls.
- Low background enables a dynamic range for complicated samples
- Suitable for Western blots, dot-blot assays, and other protein or nucleic acid methods
- 100% pure nitrocellulose
- High affinity for proteins
- Blocking ability
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
4 m | |
Western blots, dot-blot assays, and other protein or nucleic acid methods | |
100% Pure Nitrocellulose |
20 cm | |
Roll | |
0.2 μm |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Amersham Protran Premium
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Rolls > 0.2um; Roll; 20cm x 4m
Spot an opportunity for improvement?Share a Content Correction