missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Rolls

Use for the best results with fluorescence Western blotting detection applications. Cytiva Life Sciences™ Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.

250.00 € - 452.00 €

Specifications

Length (Metric) 4 m
For Use With (Application) Western blots, dot-blot assays, and other protein or nucleic acid methods
Format Roll
Material 100% Pure Nitrocellulose
View More Specs

Products 6
Product Code Brand Width (Metric) Pore Size Price Quantity & Availability  
Product Code Brand Width (Metric) Pore Size Price Quantity & Availability  
15279794
View Documents
Cytiva
10600004
30 cm 0.2 μm
452.00 €
4m
In Stock
to see stock.
Edge
Add to basket
 
15269794
View Documents
Cytiva
10600003
30 cm 0.45 μm
379.00 €
0.30kg
In Stock
to see stock.
Edge
Add to basket
 
15219804
View Documents
Cytiva
10600008
20 cm 0.45 μm
332.00 €
1 roll
In Stock
to see stock.
Edge
Add to basket
 
15229804
View Documents
Cytiva
10600009
20 cm 0.2 μm
336.00 €
1 roll
In Stock
to see stock.
Add to basket
 
15269804
View Documents
Cytiva
10600013
150 mm 0.45 μm
250.00 €
1 roll
In Stock
to see stock.
Add to basket
 
15279804
View Documents
Cytiva
10600014
150 mm 0.2 μm
273.00 €
1 roll
In Stock
to see stock.
Add to basket
 
Description

Description

Supplied in rolls.

  • Low background enables a dynamic range for complicated samples
  • Suitable for Western blots, dot-blot assays, and other protein or nucleic acid methods
  • 100% pure nitrocellulose
  • High affinity for proteins
  • Blocking ability

Specifications

Specifications

4 m
Western blots, dot-blot assays, and other protein or nucleic acid methods
Roll
100% Pure Nitrocellulose
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

Supplemental information
  • MIXTURE LIST-Contains : Nitrocellulose

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.